Learn More
Four elements solution - Ca, Mg, K, Na for ICP/MS 1g/L HNO3 matrix, Fisher chemical™
Product Code. 12916203 Shop All Fisher Chemical Products
Click to view available options
Quantity:
50 mL
100 mL
250 mL
Unit Size:
100mL
250mL
50mL
Product Code. 12916203
Brand: Fisher Chemical MS 010 250
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Quantity 250 mL