Learn More
Poulten Graf™ Fortuna™ Optima™ All Glass Luer-Lock Syringe
Able to be autoclaved at 134°C. Poulten Graf™ Fortuna™ Optima™ All Glass Luer-Lock Syringes come with an interchangeable piston.
Brand:  Poulten Graf™ 7 14021
 View more versions of this product
Product Code. 10262151
  • 10.50€ / Each
Explore more special offers
Description
Description
  • Complies with CE EN 46001 standards
  • Constructed from Durobax™ glass
  • With permanent amber graduation
  • Acid and alkali resistant 
Specifications
Specifications
Autoclavable
Borosilicate Glass
All Glass Luer-lock Syringe
1/Ea.
Luer Lock
CE EN 46001
Amber graduation
0.05 mL
1 mL
Yes