Learn More
Foamtec Stainless Steel Corrosion Remediation Kits
Enable technicians to quickly remediate corrosion using a polishing and cleaning process that is fully compatible with cleanroom operations
Brand:  Foamtec HT4500-SSC2
Encompass
 View more versions of this product
Product Code. 15183504
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
Kits include polishing and cleaning products along with instructional videos and documentation that result in mirror smooth 304 and 316 SS surfaces that resist subsequent corrosion.
Kit Contents
19-304-555: Stainless Steel Corrosion Remediation Starter Kit for Large Surfaces
2 Handles, 10 Diamond ScrubDISKS (5 girts), 1 Silicon-Carbide ScrubDISK, 1PilloWIPE, 4 Diamond ScrubPADS (4 girts), 1 Silicon-Carbide ScrubPAD, ScrubWRIGHT Pen with 2 Belts, 25 MiraWIPE Wipers, 2 UltraSOLV Sponge, and Flash Drive with instructional Videos
19-304-556: Stainless Steel Corrosion Remediation Starter Kit
1 PilloWIPE, 4 Diamond ScrubPADS (4 girts), 1 Silicon-Carbide ScrubPAD, ScrubWRIGHT Pen with 2 Belts, 10 MiraWIPE Wipers, 1 UltraSOLV Sponge, and Flash Drive with instructional Videos
Specifications
Specifications
Small Surface Starter Kit
Cleaning corrosion on stainless steel surfaces
Sahara corrosion cleaning system
280 Diamond grit ScrubPAD, 360 Diamond grit ScrubPAD, 1350 Diamond grit ScrubPAD, 1500 Silicon Carbide grit ScrubPAD, 2000 Diamond grit ScrubPAD, 280 Diamond grit ScrubDISK™, 360 Diamond grit ScrubDISK™, FTP