Learn More
Novus Biologicals™ FNDC3B Overexpression Lysate
Brand:  Novus Biologicals™ NBP2-09565
Product Code. 18058409
  • 783.00€ / 0.10mg
Explore more special offers
Description
Description
The FNDC3B Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_1128567. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot
RIPA buffer
Homo sapiens FNDC3B, mRNA.
Native