missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Flucloxacillin, sodium salt, 95%, Thermo Scientific Chemicals

Product Code. p-7077619 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

Product Code. 15575196

Brand: Thermo Scientific Chemicals 456470010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1847-24-1
Molecular Formula C19H16ClFN3NaO5S
Molecular Weight (g/mol) 475.85
MDL Number MFCD01682108
InChI Key OTEANHMVDHZOPB-SLINCCQESA-M
PubChem CID 101643208
ChEBI CHEBI:31615
SMILES [Na+].CC1=C(C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C(=NO1)C1=C(F)C=CC=C1Cl

Specifications

Type Flucloxacillin Sodium Salt
Melting Point 176.0°C to 178.0°C
Infrared Spectrum Authentic
Loss on Drying 4.5% max.
Specific Rotation +158° to +168° (20°C, 589nm) (c=1, water) On dry substance
Quantity 1 g
Flash Point >110°C
Boiling Point 677.0°C
IUPAC Name sodium;N-[(2S,5R,6R)-2-carboxy-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptan-6-yl]-3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-carboximidate
Formula Weight 475.86
Percent Purity 98%
Chemical Name or Material Flucloxacillin, sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • Flucloxacillin, sodium salt
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.