missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Floor Stand

Floor Stand designed to warn for hazardous areas, putting valuable safety information directly at the point of need.

Brand:  Brady™ 256281

 View more versions of this product

Product Code. 11706665

  • 78.20€ / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • These floor signs are manufactured from polyester, with a clear matt anti-slip laminate to prevent falling accidents while protecting the graphics underneath.
Specifications

Specifications

HEAVY DUTY FLOORSTAND ZONE DE TRAVAIL
25.98 in.
Rectangle
450 mm
Polypropylene
ATTENTION ZONE DE TRAVAIL
660 mm
17.72 in.
Black, Red on Yellow
Floor Stand
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.