Learn More
Key Instruments™ Flo Rite™ FR4000 Model Clear Acrylic Flowmeter with Standard Inlet Valve
Brand:  Key Instruments™ FR4A43BVBN
 View more versions of this product
Product Code. 11988034
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Specifications
Flowmeter
3 to 30 LPM
100 psig
Air sampling equipment aquaculture, Desalinization equipment, Gas analyzers, Medical systems, Photoprocessing equipment, Water treatment and distribution systems
Acrylic
±0.5%
Full scale
65°C
±3%
Stainless Steel
1/8 in. NPT (F)
165 mm
FR4000
34.7 mm
Buna-N
Standard inlet valve
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products