Learn More
Key Instruments™ Flo Rite™ FR4000 Model Clear Acrylic Flowmeter with Standard Inlet Valve
Brand:  Key Instruments™ FR4A42BVBN
 View more versions of this product
Product Code. 11978034
Quantity Price
1 149.81€ / Each
10 142.00€ / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Specifications
Flowmeter
2 to 20 LPM
100 psig
Air sampling equipment aquaculture, Desalinization equipment, Gas analyzers, Medical systems, Photoprocessing equipment, Water treatment and distribution systems
Acrylic
±0.5%
Full scale
65°C
±3%
Stainless Steel
1/8 in. NPT (F)
165 mm
FR4000
34.7 mm
Buna-N
Standard inlet valve
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products