Learn More
Abnova™ FLI1 Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
Brand: Abnova™ FS0058.100uL
This item is not returnable.
View return policy
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
EWSR2,SIC-1 | |
2313 | |
100 μL |
Tube | |
Friend leukemia virus integration 1 | |
FLI1 | |
DNA/RNA | |
Liquid |