Learn More
Copan Italia™ Flexible Inoculation Loop
Used for problem free planting and streaking of cultures.
Brand:  Copan Italia™ 179CS20
Product Code. 11766538
  • 323.00€ / Pack of 4000
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • No rough plastic edges, flashing or burrs on the loop head helps problem free planting and streaking of cultures
  • Loops do not cut or gouge the agar surface during streaking
Specifications
Specifications
Single-use
Flexible soft plastic
Inoculating Loop
10 μL
Light blue
4000