missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Euro Protection™ Fleece Hat
Euro Protection™ Fleece Hat is constructed of thick, 300g/m2 polar fleece for warmth.
Brand: Euro Protection™ 5COVEB
This item is not returnable.
View return policy
Description
- Constructed of 100% polyester polar fleece,300g/m2
- Includes black tightening cord for secure fit
Specifications
Hat | |
Navy | |
Cord for tightening |
Polar Fleece | |
300 gsm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction