Learn More
Lenz Laborglasinstrumente™ Flat Flange Lid with Center Neck, Three Angled Side Necks, and One Vertical Side Neck
Brand:  Lenz Laborglasinstrumente™ 06105564
 View more versions of this product
Product Code. 11572364
  • 979.00€ / Each
Estimated Shipment: 30-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Borosilicate Glass, Duran Glass
Laboratory Flange 200
Center Neck (NS): 45/40
Reaction vessels
Flange lid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Flat Flange Lid with Center Neck, Three Angled Side Necks, and One Vertical Side Neck > Includes Laboratory Flange 200; Closure Size: Center Neck (NS): 45/40