Learn More
Dostmann Electronic™ Flash III Infrared Thermometer
Dostmann Electronic™ Flash III Infrared Thermometer is a two-in-one thermometer featuring infrared and probe measuring options.
Brand:  Dostmann Electronic™ 5020-0556
Product Code. 10293882
  • 79.95€ / Each
Explore more special offers
Récupel-bijdrage uitgesloten
Description
Description
  • Temperature measurement via contact (probe) or non-contact (infrared) methods
  • Display of current, highest or lowest temperature
  • HACCP-LED indicates HI/LO-Level for food industry measurements
  • Waterproof IP 65
  • MAX-MIN-HOLD-LOCK-function
  • °C/°F-switchable
  • LED spot size indicator
  • Clip for fixing, bag, and calibration certificate included
Specifications
Specifications
0.01 to 1.00
Flash 3 Infrared Thermometer
-33°C (Infrared), -55°C (Insertion Probe)
Infrared: -33°C to 0°C ±1.0°C to ±0.1/1°C 0°C to +65°C +
0.1°C
220°C
-33°C to +220°C (Infrared), -55°C to +220°C (Insertion Probe)