Learn More
SCAT™ Fitting with Ferrule for HPLC Systems
Product Code. 15491793 Shop All SCAT Europe Products
Click to view available options
Diameter (Metric) Outer:
1.6 mm
3.2 mm
Color:
Black
Colorless
White
Unit Size:
Pack of 5
Product Code. 15491793
Brand: SCAT™ 107048
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Connect your HPLC capillaries safely and tightly to your SCAT™ system. SCAT™ PFA fittings are chemically resistant and fit Safety Caps and Safety Waste Caps. Available for different capillary sizes.
SCAT™ fittings are available in the following versions:
  • PFA fitting with integrated ferrule in various colors and diameters
  • Tube connectors in various diameters
Specifications
Type Fitting
Color Colorless
Diameter (Metric) Outer 1.6 mm
For Use With (Application) Solvent Handling
For Use With Capillary (1.6 mm OD)
Material PFA