Learn More
Fisherbrand™ Screw Caps for Disposable Glass Tubes
Shop All Fisherbrand Products
Click to view available options
Color:
Black
Closure Size:
13-415
15-415
18-415
Material:
Phenolic
Unit Size:
Pack of 1000
Specifications
Specifications
| Closure Size | 18-415 |
| Liner Material | Rubber |
| Pre-sterilized | No |
| Product Type | Screw Cap |
| Autoclavable | Autoclavable |
| Material | Phenolic |
| Color | Black |
| Sterility | Non-sterile |
| For Use With (Equipment) | 14-959-37A,14-957-76E, 14-959-37C |