Learn More
Fisherbrand™ 96-Well Polystyrene Plates
Product Code. 13535440 Shop All Fisherbrand Products
Click to view available options
Color:
Black
Clear
White
Well Shape:
Flat Bottom (F-well)
Round Bottom (U-well)
Unit Size:
Pack of 180
Pack of 80
Product Code. 13535440
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Ideal for antibiotic screens, cell-based assays and screening compounds.
Ideal for antibiotic screens, cell-based assays and screening compounds
Designed for compatibility with automated equipment
  • Polystyrene
  • Plates stack easily without lids
  • Available in flat or round well shapes
  • Nonsterile
  • Untreated, provides uniform surface properties
  • Use 12-565-511 lid (sold separately)
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
Color Black
Material Polystyrene
Volume (Metric) Well 400 μL
No. of Wells 96
Volume (Metric) Working 50-250 μL/well
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
For Use With (Application) Ideal for antibiotic screens, cell based assays and screening compounds
Quantity 180/Cs.