Learn More
Fisherbrand™ Notched Glass Plate for Owl™ P10DS Dual Gel System
Product Code. 11721586 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Owl P10DS Dual Gel System
Unit Size:
Each
Product Code. 11721586
Brand: Fisherbrand™ P1020R
Only null left
Add to basket
Add to basket
Notched Glass Plate for Owl™ P10DS Dual Gel System
  • For use with Owl P10DS dual gel system
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl P10DS Dual Gel System
Type Notched Glass Plate
Material Glass
Length (English) Gel 7.6 in.
Length (Metric) 20 cm
Width (English) Gel 7.8 in.
Width (Metric) 20 cm
Width (Metric) Gel 20 cm
Height (English) 0.12 in.
Height (Metric) 0.32 cm