Learn More
Fisherbrand™ Glass Plates for FB-VE16-1 Vertical Electrophoresis System
Product Code. 11933035 Shop All Thermo Scientific Products
Click to view available options
Type:
Blank Glass Plate
Notched Glass Plate
Offset Glass Plate
Unit Size:
Each
Product Code. 11933035
Brand: Fisherbrand™ P112G
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Different styles of glass plates to choose from.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl P1-CST Multiple Gradient Gel Caster
Type Offset Glass Plate
Material Glass
Thickness (English) 0.12 in.
Thickness (Metric) 0.3 cm
Length (Metric) 16 cm
Width (Metric) 14 cm
Height (English) 0.12 in.
Height (Metric) 0.32 cm