Learn More
Fisherbrand™ Comb for use with FB-VE10-1 Vertical Electrophoresis System
Product Code. 12404667
Click to view available options
Thickness (Metric) Teeth:
0.05 cm
0.08 cm
0.15 cm
0.77 cm
Teeth Count:
6
8
10
12
15
20
Unit Size:
Each
Product Code. 12404667
Brand: Fisherbrand™ MP8A
For use with Fisherbrand™ FB-VE10-1 Vertical Electrophoresis System.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl P8 Series Electrophoresis System
Type Comb
Gel Dimensions 10 x 10 cm
Teeth Count 8
Thickness (Metric) Teeth 0.77 cm
Volume (Metric) Well 62 μL
Length (English) Footprint 5.9 in.
Length (English) Gel 3.9 in.
Length (Metric) Footprint 15 cm
Length (Metric) Gel 10 cm
Width (English) Gel 3.9 in.
Width (English) Teeth 0.3 in.
Width (Metric) Footprint 13 cm
Width (Metric) Gel 10 cm
Width (Metric) Teeth 0.77 cm
Height (English) Footprint 5.11 in.
Height (Metric) Footprint 13 cm