Learn More
Fisherbrand™ Traceable™ Digital Thermometer with Stainless-Steel Stem, 0.25 in. LCD Screen, and Protective Guard
Product Code. 11749715 Shop All Fisherbrand Products
Click to view available options
Temperature Range (Metric):
-50°C to +150°C
-50°C to +250°C
-50°C to +280°C
-50°C to +300°C
Temperature Range (English):
-58°F to +302°F
-58°F to +482°F
-58°F to +536°F
-58°F to +572°F
Accuracy:
±0.2°C
±0.4°C
±0.5°C
±1°C
±1.5°C
Length (Metric) Probe:
8.9 cm
20 cm
20.32 cm
28.87 cm
Unit Size:
Each
Récupel-bijdrage uitgesloten
Product Code. 11749715
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
Easy-to-read handheld Traceable™ Digital Thermometer.
  • Longer probes are excellent for measuring sample temperatures while stirring in deep vessels or water baths
  • Thermometers with longer sampling rates allow thermometer to be removed from sample before reading—ideal when display is difficult to read in measuring position
  • For use in cuvets, soil, incubators, petroleum products and foods
Certifications
Multi-point calibration on individually-numbered Traceable Certificate assures accuracy from ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA; indicates traceability to NIST.
Specifications
Certifications/Compliance DNV, ISO 17025, A2LA
Diameter (English) Probe 0.14 in.
Diameter (Metric) Probe 3.56 mm
Includes Traceable certificate, Stainless steel probe
Length (English) Probe 8 in.
Length (Metric) 235 mm
Length (Metric) Probe 20.32 cm
Material Plastic
Resolution 0.1°C
Weight (English) 1 oz.
Weight (Metric) 20 g
Product Type Digital Thermometer
Length (English) 9.25 in.
Max. Temperature (English) 536°F
Max. Temperature (Metric) 280°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58°F to +536°F
Temperature Range (Metric) -50°C to +280°C
Calibrated Yes
For Use With (Equipment) Cuvettes, soil, incubators, petroleum products and foods, wet labs, wash down areas and field use
Accuracy ±1°C
Display Digital LCD
Width (Metric) 8 cm