Learn More
Thermo Scientific™ Finnpipette™ Stands
Provide safe and convenient storage for manual and electronic pipettes using these stands, with options for single stand, F-stand, multi-stand, multichannel and carousel.
16.15€ - 223.00€
Product Code | Brand | Description | Holds | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Holds | Price | Quantity & Availability | ||||
11455748
|
Thermo Scientific™
9420400 |
F-Stand, Linear Stand, white | Manual Pipettes (6) |
147.00€
Each |
In Stock
Log in to see stock. |
||||
12396512
|
Thermo Scientific™
9420030 |
Multichannel Single Pipette Multiposition Stand | N/A | ||||||
11769855
|
Thermo Scientific™
2206040 |
Finnpipette Shelf Hanger | Manual Pipette (1) |
16.15€
Each |
Estimated Shipment: 04-02-2025
Log in to see stock. |
||||
11478419
|
Thermo Scientific™
2209480 |
3 |
20.75€
1 set |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
11488419
|
Thermo Scientific™
2209490 |
Finnpipette Novus Adapter for Carousel Stand | 1 | N/A | |||||
10319080
|
Thermo Scientific™
9420290 |
Pipette Stand, gray | Manual Pipettes (6) |
141.00€
Each |
In Stock
Log in to see stock. |
||||
11418419
|
Thermo Scientific™
9420320 |
Pipette Mini-Stand, gray | Manual Pipettes (3) |
109.00€
Each |
In Stock
Log in to see stock. |
||||
11716135
|
Thermo Scientific™
9420340 |
Pipettes (6 Manual or 3 Electronic) |
223.00€
Each |
In Stock
Log in to see stock. |
|||||
11726924
|
Thermo Scientific™
9420360 |
Electronic Pipette (1) |
110.00€
Each |
In Stock
Log in to see stock. |
|||||
11817391
|
Thermo Scientific™
9420390 |
Multichannel Single Pipette Stand | Multichannel Manual Pipette (1) |
27.25€
Each |
In Stock
Log in to see stock. |
||||
Description
Provide safe and convenient storage for manual and electronic pipettes using Thermo Scientific™ Finnpipette™ Stands. Available options include options for single stand, F-stand, multi-stand, multichannel and carousel.
Finnpipette F-Stand
- Accommodates six pipettes
- Sturdy crossbar structure with slot for each pipette for secure storage
- Suitable for both single channel and multichannel models
Finnpipette Multichannel Stand
- Multi-position stand
- Can hold any multichannel Finnpipette
- Pipette can be stored in upright or horizontal position
- Small footprint
Finnpipette Carousel Stand
- Rotating, portable
- Transparent lid protects pipettes from contamination
- Shipped with three adapters for electronic Finnpipette Novus pipettes and five adapters for manual Finnpipette pipettes
Finnpipette Novus Stand
- For convenient storage of electronic Finnpipette Novus pipette
- Accommodates both single and multichannel Finnpipette Novus models with ease