Learn More
Coverguard™ FFP2 NR D SL VO-GA Half Mask for Organic Vapors and Acids, Blue with Exhalation Valve
The blue FFP2 half masks have an electrostatic filter media to filter bad odors. The design gives limited breathing resistance, thanks to the filtering material and exhalation valve with active carbon, aids breathing and avoids fog build-up inside the mask.
Brand:  Coverguard™ 23246
354.68 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Product Code. 12043196
  • 532.00€ / Pack of 120
Estimated Shipment: 07-08-2025
to see stock.
Explore more special offers
Add to basket
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Description
  • PPE Cat. III FFP2 NR D Organic vapors, Gas and Acids
  • Filters bad odors
  • Exhalation valve with active carbon reduces fog build-up inside the mask
  • Non-woven polypropylene
  • Adjustable aluminum nose clip with PU foam total nose seal
  • Specific design gives limited breathing resistance thanks to the large surface of the filtering material
Specifications
Specifications
CE PPE Cat. III, FFP2, EN 149:2001 + A1:2009
Single-use
Heavy & Process Industries, Chemical & Pharmaceutical Industries, Light Industries, Infrastructure, Building Industry, Construction - Finishing Work, DIY
Aerosols and Low Toxic Dust
Non-sterile
Blue FFP2 Half Mask with Shell and Carbon Valve
Yes
Adjustable Aluminum Nose Clip, PU Foam Nose Pad
Half Mask
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Coverguard™ FFP2 NR D SL VO-GA Half Mask for Organic Vapors and Acids, Blue with Exhalation Valve > Face Mask; Nosepiece Type: Adjustable Aluminium Nose Clip, PU Foam Nose Pad