Learn More
Coverguard™ FFP2 NR D SL VO Face Mask for Organic Vapors Gas and Acids with Exhalation Valve
FFP2 half masks have an electrostatic filter media to filter bad odors. The design gives limited breathing resistance, thanks to the filtering material and exhalation valve with active carbon, aids breathing and avoids fog build-up inside the mask.
Brand:  Coverguard™ 23238
Product Code. 12033196
  • 405.00€ / Pack of 120
Estimated Shipment: 04-04-2025
to see stock.
Explore more special offers
Description
  • PPE Cat. III FFP2 NR D Organic vapors, Gas and Acids
  • Filters bad odors
  • Exhalation valve with active carbon reduces fog build-up inside the mask
  • Non-woven polypropylene
  • Adjustable aluminum nose clip with PU foam total nose seal
  • Specific design gives limited breathing resistance thanks to the large surface of the filtering material
Specifications
Specifications
CE PPE Cat. III, FFP2 NR D, EN 149:2001 + A1:2009
Single-use
Heavy & Process Industries, Chemical & Pharmaceutical Industries, Light Industries, Infrastructure, Building Industry, Construction - Finishing Work, DIY
Aerosols and Low Toxic Dust, Organic Vapors, Gas and Acids
Non-sterile
FFP2 Half Mask with Shell and Carbon Valve
Yes
Aluminum Adjustable Nose Clip, PU Foam Nose Pad
Half Mask