Learn More
Coverguard™ FFP2 NR D SL Half Mask with Exhalation Valve
The preformed design of the Disposable FFP2 NR D SL Half Mask fits with glasses or goggles to avoid the risk of condensation. The ergonomic form offers an optimal adjustment to the face, without a nasal clip. Breathing resistance is reduced with the exhalation valve.
Brand: Coverguard™ 23276
This item is not returnable.
View return policy
Description
- FFP2 NR D PPE Category III
- Preformed design fits with glasses or goggles
- Ergonomic form offers optimal fit
- Exhalation valve reduces breathing resistance
Specifications
CE PPE Cat. III, FFP2 NR D, EN 149:2001 + A1:2009 | |
Single-use | |
Aerosols and Low Toxic Dust |
FFP2 Preformed Shell Mask with Valve | |
Yes | |
Non-sterile |