Learn More
Coverguard™ FFP2 NR D Face Mask
The Coverguard™ FFP2 masks are single-use and protect against non-toxic solid and liquid aerosols in applications such as farming, silviculture and fishing, light industries, maintenance, infrastructure and building, DIY, as well as general use in the lab or field.
219.00€ - 696.00€
Specifications
Certifications/Compliance (Europe) CE PPE Category III FFP2 NR D, EN 149:2001 + A1:2009
Disposable Single-use
For Use With (Equipment) Chemical and Pharmaceutical Industries, Light Industries, Heavy and Process Industries, Construction Finishings, Building Industry, Infrastructure
Resistant To Non-Toxic Solid and Liquid Aerosols
Sterility Non-sterile
View More Specs
Products 3
Product Code Brand Style Nosepiece Type With Exhalation Valve Price Quantity & Availability  
Product Code Brand Style Nosepiece Type With Exhalation Valve Price Quantity & Availability  
12073186
View Documents
Coverguard™
23201
Shell Mask Adjustable Nose Clip with PU Foam Nose Pad No
511.00€
Pack of 240
Estimated Shipment: 30-12-2024
to see stock.
Add to basket
12083186
View Documents
Coverguard™
23205
Half Mask with Exhalation Valve Adjustable Nose Clip Yes
696.00€
Pack of 240
Estimated Shipment: 02-04-2025
to see stock.
Add to basket
12093186
View Documents
Coverguard™
23206
Shell Mask with Exhalation Valve Adjustable Nose Clip with PU Nose Pad Yes
219.00€
Pack of 120
Estimated Shipment: 07-03-2025
to see stock.
Add to basket
Description
Description
  • PPE Cat. III FFP2 NR D
  • Disposable
  • Electrostatic filter media
  • Sliding elastic band for better adjustment
  • Latex-free, non-woven polypropylene
  • Adjustable nose clip
Specifications
Specifications
CE PPE Category III FFP2 NR D, EN 149:2001 + A1:2009
Chemical and Pharmaceutical Industries, Light Industries, Heavy and Process Industries, Construction Finishings, Building Industry, Infrastructure
Non-sterile
Single-use
Non-Toxic Solid and Liquid Aerosols