Learn More
Coverguard™ FFP1 NR D Face Mask
The Coverguard™ FFP1 masks are single-use and protect against non-toxic solid and liquid aerosols, in applications such as farming, silviculture and fishing, light industries, maintenance, infrastructure and building, DIY, as well as general use in the lab or field.
Brand:  Coverguard™ 23100
 View more versions of this product
Product Code. 12003186
  • 503.00€ / Pack of 240
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • PPE Cat. III FFP1 NR D
  • Disposable
  • Electrostatic filter media
  • Sliding elastic band for better adjustment
  • Latex-free, non-woven polypropylene
  • Adjustable nose clip
Specifications
Specifications
CE PPE Category III FFP1 NR D, EN 149:2001 + A1:2009
Single-use
No
Yes
Adjustable Nose Clip
Non-sterile
FFP1 Disposable Foldable Half-Mask, Sealed Outer Contour
Non-woven Polypropylene
FFP1
Synthetic Rubber Elastic (Latex Free)
Non-Toxic Aerosols and Non-Toxic Dusts
Flat Foldable Half-Mask