Learn More
Corning Media Tech Fetal Bovine Serum
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
- Raw pooled serum is filtered through series of 0.1μ sterilizing grade filters
- Low endotoxin levels
- Low hemoglobin levels
Specifications
Specifications
| Form | Liquid |
| Product Type | Fetal Bovine Serum |
| Quantity | 500 mL |
| Sterility | Sterile |
| Sterilization Method | Sterile-filtered |
| Endotoxin Concentration | <20 EU/mL |
| For Use With (Application) | Cell and tissue culture |
| Hemoglobin Concentration | <30 mg/dL |
Product Content Correction
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido