Learn More
Thermo Scientific™ RNase A/T1 Mix
Get the RNA degradation activity of both RNase A and RNase T1 with Thermo Scientific™ RNase A/T1 Mix.
Brand: Thermo Scientific™ EN0551
78.67 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Thermo Scientific RNase A/T1 Mix combines the RNA degradation activity of both RNase A and RNase T1. The RNase A specifically hydrolyzes RNA at C and U residues; RNase T1 specifically hydrolyzes RNA at G residues (see Figure 1 in Supporting Data [see Reference 1]).
Highlights
- Higher level of RNA degradation than with RNase A and RNase T1 separately
- Free of DNase activity - It is not necessary to heat the mix before use
Specifications
RNase | |
1 mL | |
RNase A/T1 Mix |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Promotions
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Days
33Hours
20Minutes
6Seconds
39 -
Multiple Manufacturers
More Discovery, Less Cost
Now extended - offer ends 28 MarchPromo Code: 24111Expires: 2025-03-28
Days
0Hours
20Minutes
6Seconds
39New Offer Save Now -
Multiple Manufacturers
Grow Your Discovery
Fall in love with big savingsPromo Code: 24107Expires: 2025-03-28
Days
0Hours
20Minutes
6Seconds
39