Learn More
Thermo Scientific™ Lambda DNA/HindIII Marker, ready-to-use
Perform sizing and approximate quantification of large double-stranded DNA fragments in agarose gels with this conventional, ready-to-use marker composed of 8 sharp bands.
Brand: Thermo Scientific™ SM0103
60.16 EUR valid until 2024-12-14
Use promo code "21632" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21632” to get your promotional price
Description
The Thermo Scientific Lambda DNA/HindIII Marker, ready-to-use is recommended for sizing of linear double-stranded large DNA on agarose gels. Lambda DNA is digested to completion with the appropriate Thermo Scientific restriction enzyme(s) and purified and dissolved in storage buffer. DNA fragments contain various sticky ends depending on the restriction enzyme used for preparation of the marker.
Lambda DNA/HindIII Marker, ready-to-use is premixed with 6X DNA Loading Dye for direct loading (after heating) onto agarose gels.
Highlights
- Sizing and approximate quantification of large DNA fragments
- Sharp bands
- Premixed with 6X DNA Loading Dye for a direct loading (after heating) and for room temperature storage
- Supplied with loading dye for sample DNA
Specifications
6X | |
DNA/Hind III Marker | |
Agarose Gels |
Agarose | |
Yes | |
5 x 50 μg |
For Research Use Only. Not for use in diagnostic procedures.