Learn More
Fenhexamid, Honeywell Fluka™
Brand:  Honeywell Fluka R1740-100MG
Product Code. 16684534
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.
  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fenhexamid
1
N-(2,3-Dichloro-4-hydroxyphenyl)-1-methylcyclohexanecarboxamide
1.34 g/cm3 (20°C)
Beige
>150°C
126833-17-8
8996658
302.20 g/mol
155°C