Learn More
Fc gamma RIIB/CD32b Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-99136APC
Product Code. 18616315
  • 430.00€ / 0.10mL
Description
Fc gamma RIIB/CD32b Polyclonal antibody specifically detects Fc gamma RIIB/CD32b in Human samples. It is validated for ELISA