Learn More
MP Biomedicals™ FastPrep-24, Second Year Extended Limited Warranty
Product Code. 15252810 Shop All MP Biomedicals Products
Click to view available options
Unit Size:
Each
Product Code. 15252810
Brand: MP Biomedicals™ 116002599
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Warranty can be purchased anytime before the one year new instrument standard manufacturer's warranty expires.
Covers manufacturer's defects and includes repair or replacement of such defects, including transportation of instrument to be repaired or replaced without charges. Does not cover normal wear and tear or replacement of electrical cord, springs, rubber cups, dome, tubes holders, tube convers or other accessories.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) FastPrep 24 Classic