missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fast Green FCF, pure, certified

Product Code. 11906941
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2353-45-9
Molecular Formula C37H34N2Na2O10S3
Molecular Weight (g/mol) 808.84
MDL Number MFCD00013053
InChI Key XJBPDZVCYTYRIN-UHFFFAOYSA-L
Synonym C.I. 42053
PubChem CID 16887
IUPAC Name disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-5-hydroxybenzenesulfonate
SMILES [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O

Specifications

Melting Point 290.0°C
Absorption Ratio 0.98 to 1.2
Dye Content 85% min.
Solubility Information Solubility in water: 150g/L (25°C).
Infrared Spectrum Authentic
Lambda Max 622 to 626nm
Formula Weight 808.84
Grade Pure
Packaging Glass bottle
Color Bordeaux to Maroon
Quantity 100 g
Physical Form Powder
Chemical Name or Material Fast Green FCF
Show More Show Less
compliance-icons
Product Identifier
  • Fast green FCF
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.