missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ 96-Well, Cell Culture-Treated, Angled-Bottom, Insert Plate Microplate
GREENER_CHOICE

Color:Clear; Sterility:Sterile; Lid:With Lid

Brand:  Corning 353938

Product Code. 10520653

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Automation compatible design
  • Format compatible with most robots and fluid handling instruments
  • Complete sample recovery
  • Features an angled bottom for more complete sample utilization
  • Bottom plate used in conjunction with the insert for culturing cells eliminates the transfer step from single-well feeder tray for sample analysis
  • Excellent reproducibility
  • Total assay flexibility—ideal for transport studies
Storage: room temperature, away from direct sunlight
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
0.08 cm2
1 μm
Polystyrene
Sterile
5 insert plates with 96 square well angled-bottom plates; lids
5
3.2 mm
With Lid
8 x 12
Clear
96
Cell Culture
5
5/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.