Description
- Soft, crystal-clear PVC frame goggles with a wide support area all around the face to enhance comfort.
- Specific channel on the side of the frame to accommodate the temple of prescription frames.
- Model fits well with prescription glasses.
- Special neoprene headband for good chemical resistance and easy cleaning.
- Specifically designed to carry tear-off folies.
- One replacement strip included.
- Anti-Fog coating to help reduce lens fogging.
Specifications
Specifications
Lens Coating | Anti-fog |
Lens Tint | Clear |
Product Type | Safety Goggles |
For Use With (Application) | Construction, food industry |
Headband Material | Neoprene |
No. per Pack | 10/Pk. |
Ventilation Details | Indirect ventilation |
Material | PVC |
Lens Material | Acetate |
Frame Type | Soft Frame |
Product Suggestions
Customers who viewed this item also viewed
Documents
Product Information
Datasheets
7000062783
File size: 217 kB
Product Content Correction
Product Title
3M™ Fahrenheit™ Safety Goggles >