missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ F1.5 Model ThermoMixer™

Use this thermal cycler for transformation of bacteria strains and plasmids. Eppendorf™ F1.5 Model ThermoMixer™ have a fixed block for 0.5/1.5/2.0 mL tubes and plates. Prevention of condensate formation in small vessels can be achieved with the heated lid Eppendorf ThermoTop (optional).

Brand:  Eppendorf™ 5385000016

 View more versions of this product

Product Code. 13508470

  • 3310.00€ / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Anti-spill technology prevents lid wetting and cross-contamination
  • Heating and mixing in 0.5/1.5/2.0 mL reaction vessels (F0.5/F1.5/F2.0) or plates (FP)
  • Up to 1,500rpm (F1.5/F2.0) or 2,000rpm (F0.5/FP) for efficient sample mixing
  • Outstanding 2DMix-Control technology results in excellent mixing performance
  • Predefined temperature keys (37°C, 42°C, 56°C and 95°C) for simple and intuitive operation
  • Very fast heat up speed of up to 18°C/min, reducing your waiting time
  • Eppendorf PhysioCare Concept provides ergonomic design and operation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Max. ±0.5°C at 20°C to 45°C
With Plate Smartblock
220 to 240 V, 50/60 Hz, 200 W
170 mm
304 mm
50 to 60 Hz
300 to 1500 rpm
1 to 100°C
220 to 240 V
ThermoMixer F1.5
206 mm
Used to Transformation of Bacteria Strains and Plasmids
USB Interface
1500 rpm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.