missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ F(ab')2-Goat anti-Human IgG Fc Secondary Antibody, FITC
F(ab')2-Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ H10101C
This item is not returnable.
View return policy
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Human IgG Fc | |
| Polyclonal | |
| FITC | |
| Assay-dependent | |
| Goat | |
| 1 mL | |
| Secondary | |
| 0.1% Sodium azide | |
| 4°C, store in dark | |
| Liquid | |
| HRP, AP, fluorescence |
| Flow Cytometry, Immunocytochemistry | |
| 0.7mg/mL | |
| F(ab')2-Goat anti-Human IgG Fc Secondary Antibody, 1mL, 0.7mg/mL Concentration, Liquid, Goat Host, Human Species Reactivity, FITC Conjugate, IgG Isotype, 4°C Storage Temperature, PBS with BSA Storage Buffer, For Flow Cytometry | |
| PBS with BSA and 0.1% sodium azide | |
| Purified | |
| ASR, RUO | |
| 1 Each | |
| Human | |
| Secondary Antibody | |
| IgG | |
| Responsibly packaged |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction