missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ eZKine™ CD8 Activation 1 Whole Blood Intracellular Cytokine Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
25 tests
Description
Includes
- eZKine CD8 Activation 1 Cocktail : 25 tests. This cocktail contains the following antibodies:
- Anti-Human CD3 (SK7) APC
- Anti-Human CD8 (SK1) PerCP-eFluor 710
- Anti-Human CD69 (FN50) PE
- Anti-Human IFN gamma (4S.B3) FITC
- eZKine Isotype Control Cocktail D : 25 tests. This cocktail contains the following antibodies:
- Anti-Human CD3 (SK7) APC
- Anti-Human CD8 (SK1) PerCP-eFluor 710
- Mouse IgG1 K Isotype Control PE
- Mouse IgG1 K Isotype Control FITC
- eZKine Fix/Lyse Concentrate (4X)
- eZKine Fix/Lyse Diluent
- Permeabilization Buffer (10X)
- Stimulation of whole blood in the presence of Protein Transport Inhibitor Cocktail (cat. 00-4980, not included) is followed by red blood cell lysis and fixation in a single step and subsequent permeabilization.
- Samples are then ready to stain with the CD8 Activation 1 cocktail and concentration-matched Isotype Control cocktail.
Specifications
Specifications
| Reactivity | Human |
| Detects | CD3, CD69, CD8, IFN gamma |
| For Use With (Application) | Intracellular Staining Followed by Flow Cytometric Analysis |
| Quantity | 1 Kit |
| Content And Storage | 2°C to 8°C. Do not freeze. Light sensitive material. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction