missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ EZ-Link™ TFP Ester-PEG4-DBCO
An amine reactive heterobifunctional crosslinker which contains a dibenzylcyclooctyne (DBCO) group for copper-free click chemistry applications. The crosslinker can be used to label a variety of amine containing molecules such as proteins, biomolecules, or small molecules.
222.00€ - 301.00€
Specifications
Chemical Reactivity | Amine-Azide |
---|---|
Cleavable | No |
Description | TFP Ester-PEG4-DBCO |
Molecular Weight (g/mol) | 714.7 |
PEGylated | Yes |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17514346
|
Thermo Scientific™
C20039 |
25 mg |
222.00€
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17621586
|
Thermo Scientific™
C20043 |
10 x 1 mg |
301.00€
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- An amine reactive heterobifunctional crosslinker which contains a dibenzylcyclooctyne (DBCO) group for copper-free click chemistry applications
- The crosslinker can be used to label a variety of amine containing molecules such as proteins, biomolecules, or small molecules
- Once a protein or (bio)molecule is DBCO labeled, it can undergo a copper-free azide-alkyne cycloaddition reaction to form a stable triazole linkage with an azide labeled coupling partner
- Can be used to create diverse conjugates with the biorthogonal coupling between an alkyne and azide (supplied separately)
- The coupling or crosslinking procedure is typically done in two steps
- First, a protein or amine containing molecule is labeled with a DBCO group and excess crosslinker is removed
- Second, an azide containing moiety (prepared independently) is introduced into the same system as the DBCO moiety
- Labeling occurs under biological conditions without the introduction of any additional reagents such as a copper catalyst
- Ragent readily dissolves in water-miscible solvents for dilution into aqueous reaction mixtures
- TFP esters are more hydrolytically stable than NHS esters
Specifications
Amine-Azide | |
TFP Ester-PEG4-DBCO | |
Yes | |
Upon receipt store desiccated at -20°C. | |
Wet Ice | |
Chemical Labeling | |
TFP Ester, DBCO | |
DMF, DMSO | |
No |
No | |
714.7 | |
17.9 Å | |
Not determined | |
EZ-Link™ No-Weigh™ | |
Heterobifunctional | |
Medium (10 to 30 Å) | |
Standard, Single-use | |
Crosslinker |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ EZ-Link™ TFP Ester-PEG4-DBCO