Learn More
Thermo Scientific™ EZ-Link™ Sulfo-NHS-SS-Biotin, No Weigh Format
Product Code. p-4531571 Shop All Thermo Scientific Products
Click to view available options
Quantity:
8 x 1 mg
100 mg
10 x 1 mg
Unit Size:
100mg
10mg
Pack of 8
Product Code. 10361554
Brand: Thermo Scientific™ 21328
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Thermo Scientific™ EZ-Link™ Sulfo-NHS-SS-Biotin No-Weigh™ Format is a water-soluble, NHS-ester biotinylation reagent whose spacer arm includes a cleavable disulfide bond for reversible labeling of proteins and cell surface primary amines.
Thermo Scientific™ EZ-Link Sulfo-NHS-SS-Biotin enables simple and efficient biotin labeling of antibodies, proteins and any other primary amine-containing biomolecules in solution.
Highlights:
  • Cleavable analog of Sulfo-NHS-LC-Biotin
  • Can remove biotin label by cleaving spacer disulfide bond with reducing agents, such as DTT
  • Ideal for affinity purification experiments that require efficient elution from immobilized avidin supports (cleaving disulfide bond releases proteins)
  • Long spacer arm reduces steric hindrance for binding interactions
TRUSTED_SUSTAINABILITY
Specifications
Cleavable Yes
Description EZ-Link Sulfo-NHS-SS-Biotin, No-Weigh Format
Formulation Sulfo-NHS ester of biotin with an ethyl-1,3-dithioproprionate spacer
Molecular Weight (g/mol) 606.69
Membrane Permeable No
PEGylated No
Spacer Arm Length 24.3
Quantity 8 x 1 mg
Sufficient For 8 uses to label a total of 88mg of IgG using typical conditions
Content And Storage -20°C protected from moisture
Chemical Reactivity Amine
Solubility Water