Learn More
Esculape Laboratoire™ Eyewash Station
Administer emergency eye care using an Esculape™ Eyewash Station. Components of this station include those that rinse and evacuate foreign substances from the eyes.
Brand:  Esculape Laboratoire™ 8205200
 View more versions of this product
Product Code. 11769522
  • 102.00€ / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Fixed position with wall bracket;
  • Ergonimic eye cup aids continuous rinsing
  • Quick release bottle caps
  • Instructions in three languages
  • Three year shelf life if unopened
Specifications
Specifications
For Remove Particles or Dust, Rehydrates the Eye
0.9% Sodium Chloride, 500 mL
Eyewash Station
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products