missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente Extractors for Specific Light Solvents
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
511.34€ - 707.55€
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10684582
|
Lenz Laborglasintrumente
05432058 |
1 L |
707.55€
Each |
Estimated Shipment: 30-09-2024
Log in to see stock. |
|||||
10644091
|
Lenz Laborglasintrumente
05432037 |
250 mL |
511.34€
Each |
Estimated Shipment: 30-09-2024
Log in to see stock. |
|||||
10674582
|
Lenz Laborglasintrumente
05432049 |
500 mL |
564.77€
Each |
Estimated Shipment: 30-09-2024
Log in to see stock. |
|||||
Description
- Extractor head with insert filter (frit, porosity: 0)
- With PTFE valve stopcock
- Made of DURAN™ tubing
- Dimroth condenser
- With thread GL 14
- Round bottom flask
Specifications
0 |