Learn More
Memmert™ Extended Overtemperature Protection for SF30
Extended overtemperature protection with integrated Pt100 sensor for independent temperature monitoring.
Brand:  Memmert™ SF30A6
Product Code. 17508215
  • 195.00€ / Each
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory
  • With integrated Pt100 sensor
  • For models with SingleDISPLAY
Specifications
Specifications
SF30 Sterilizer
Integrated Pt100 Sensor