Learn More
Novus Biologicals™ Exosome Standards (SK-N-SH cell line)
Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications
Brand: Novus Biologicals™ NBP2-49852-600ug
1076.00 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Store at 4°C. After reconstitution store at -70°C. | |
LYOPH | |
6 x 100 μg Vials |