missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ EV MaxiSEC Columns
Shop All Bio Techne Products
missing translation for 'orderingAttributeHoverText'
Quantity:
3 Columns
missing translation for 'unitSize'
Pack of 3
Specifications
Specifications
| Quantity | 3 Columns |
| Content And Storage | Store at 4°C. |
| Sample Type | Urine, cell culture supernatant, or other diluted matrices |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction