missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ Salinity (NaCl) Standard Solutions

Product Code. 16445339
Click to view available options
Concentration:
25 ppt
5 ppt
Unit Size:
480mL
Each
This item is not returnable. View return policy

Product Code. 16445339

Brand: Thermo Scientific™ 01X211231

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Eutech™ offers Salinity Standard Solutions for use with Salinity Meters.

Eutech™ offers a range of Salinity Standard Solutions for use with Salinity Meters. Salinity measurements are common in industries like agriculture, aquaculture, hydroponics, food, pools and spas where it is necessary to monitor the salt level constantly.

Features:

  • Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.

Description:

  • ECNACL5PPT: 5 ppt (parts per thousand) Nacl calibration solution
  • ECNACL25PPT: 25 ppt (parts per thousand) Nacl calibration solution
  • ECNACL45PPT: 45 ppt(parts per thousand) Nacl calibration solution
TRUSTED_SUSTAINABILITY

Specifications

Concentration 25 ppt
Description 25 ppt NaCl Calibration Solution
Type NaCl Calibration Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.