missing translation for 'onlineSavingsMsg'
Learn More

Eutech™ KCl Calibration Solution

For conductivity calibration and standards, choose from our available Eutech™'s Conductivity Standards, Calibrations, and Solutions

Brand:  Thermo Scientific Eutech 01X211207

 View more versions of this product

Product Code. 10775193

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Eutech™ KCl Calibration Solutions are used for calibrating conductivity probes for accurate measurement. Get quick and accurate conductivity measurements using Eutech™ conductivity meters and probes in the field, laboratories, and schools.

Features:

  • Packed in 60mL or 480 ml durable plastic bottles
  • Economical and convenient to use
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1413 μS/cm KCl calibration solution
Calibration Solution
Conductivity Standard
7
1413 μS/cm
480 mL
Colorless
Conductivity Meters and Electrodes
102.4°C
Economical and Convenient for Laboratory Use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions