missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Europium Mini Beads Antibody Conjugation Kit
Novus Biologicals™ Europium Mini Antibody Labeling Kit allows antibodies or proteins to quickly and easily conjugated to 200nm Europium chelate microspheres.
1506.17€ - 2830.85€
Specifications
Content And Storage | -20°C |
---|---|
Kit Contents | 4 or 10 Mini vials of 200nm Europium (dependent on kit), 1 vial of 1x Reaction Buffer A, 1 vial of 1x Reaction Buffer B, 1 vial of 10x Europium Quencher, 1 vial of 1x Resuspension Buffer |
Product Type | Europium Mini Antibody Labeling Kit |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
18003826
|
Novus Biologicals™
1200-0100 |
10 reactions |
2830.85€
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18006204
|
Novus Biologicals™
1200-0040 |
4 reactions |
1506.17€
16µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kit Components:- 4 or 10 Mini vials of 200nm Europium (dependent on kit)
- 1 vial of 1 x Reaction Buffer A
- 1 vial of 1 x Reaction Buffer B
- 1 vial of 10x Europium Quencher
- 1 vial of 1 x Resuspension Buffer
Specifications
-20°C | |
4 or 10 Mini vials of 200nm Europium (dependent on kit), 1 vial of 1x Reaction Buffer A, 1 vial of 1x Reaction Buffer B, 1 vial of 10x Europium Quencher, 1 vial of 1x Resuspension Buffer | |
Europium Mini Antibody Labeling Kit |
For research use only.