missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETI Process Technique™ Cooler And Freezer
Brand: ETI Process Technique 9600000460
This item is not returnable.
View return policy
Specifications
Cooler and Freezer | |
Gray | |
2 years | |
30 x 46.5 x 41.4 cm | |
41.4 cm | |
11.5 kg |
18 L | |
Handle | |
CDF 18 | |
30 cm | |
46.5 cm |