missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethosuximide, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 47.0°C to 50.0°C |
Boiling Point | 134.0°C (5.0 mmHg) |
Infrared Spectrum | Authentic |
Quantity | 5 g |
Solubility Information | Solubility in water: soluble. |
Formula Weight | 141.17 |
Percent Purity | ≥97.5% (HPLC) |
Chemical Name or Material | Ethosuximide |
Safety and Handling
Product Identifier
- Ethosuximide
- Danger
- Acute toxicity Category 4
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1A
- H241-Heating may cause a fire or explosion.
- H302-Harmful if swallowed.
- H360D-May damage the unborn child.
- P201-Obtain special instructions before use.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : 3-ethyl-3-methyl-2,5-pyrrolidinedione
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'